Assay Details
Gene Symbol: Cyp11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172939.1 | NP_001166410.1 | 604 | 68 | ||
GenBank mRNA | Z69785.1 | - | 604 | 68 |

More Information
Set Membership: |
![]() ![]() |
