Assay Details
| Gene Symbol: Guca2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172902.1 | NP_001166373.1 | 2 - 3 | 294 | 76 | |
| GenBank mRNA | Z74736.1 | - | 2 - 3 | 294 | 76 | |
More Information
Set Membership: |
|

Guca2a
Genomic Map