Assay Details
| Gene Symbol: Guca2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172958.1 | NP_001166429.1 | 1 - 2 | 127 | 92 | |
| GenBank mRNA | Z74738.1 | - | 1 - 2 | 127 | 92 | |
More Information
Set Membership: |
|

Guca2b
Genomic Map