Assay Details
Gene Symbol: Kcnj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173044.1 | NP_001166515.1 | 3 | 1443 | 100 | |
XM_005007689.3 | XP_005007746.1 | 4 | 3194 | 100 | ||
GenBank mRNA | AJ006229.1 | - | 3 | 1443 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
