Assay Details
| Gene Symbol: Cxcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172875.1 | NP_001166346.1 | 1 | 552 | 84 | |
| XM_013159993.2 | XP_013015447.1 | 2 | 623 | 84 | ||
| GenBank mRNA | AY237539.1 | - | 1 | 552 | 84 | |
More Information
Set Membership: |
|

Cxcr2
Genomic Map