Assay Details
Gene Symbol: Hsd3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172878.2 | NP_001166349.1 | 2 - 3 | 248 | 89 | |
GenBank mRNA | AY914174.1 | - | 2 - 3 | 251 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
