Assay Details
Gene Symbol: Serpinf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173060.1 | NP_001166531.1 | 5 - 6 | 712 | 82 | |
XM_013151993.1 | XP_013007447.1 | 5 - 6 | 673 | 82 | ||
GenBank mRNA | EF679792.3 | - | 5 - 6 | 712 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
