Assay Details
| Gene Symbol: Ces1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173109.1 | NP_001166580.1 | 10 - 11 | 1255 | 96 | |
| GenBank mRNA | AB010634.1 | - | 10 - 11 | 1255 | 96 | |
More Information
Set Membership: |
|

Ces1d
Genomic Map