Assay Details
Gene Symbol: Ugt2b22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173113.1 | NP_001166584.1 | 5 - 6 | 1330 | 98 | |
GenBank mRNA | AB034988.1 | - | 5 - 6 | 1330 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
