Assay Details
Gene Symbol: Ugt2b21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173115.1 | NP_001166586.1 | 1 - 2 | 723 | 81 | |
GenBank mRNA | AB034987.1 | - | 1 - 2 | 723 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
