Assay Details
| Gene Symbol: Rnasel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173055.1 | NP_001166526.1 | 1663 | 89 | ||
| GenBank mRNA | DQ497161.1 | - | 1663 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnasel| Gene Symbol: Rnasel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173055.1 | NP_001166526.1 | 1663 | 89 | ||
| GenBank mRNA | DQ497161.1 | - | 1663 | 89 | ||
Set Membership: |
|