Assay Details
Gene Symbol: Ambn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173124.1 | NP_001166595.1 | 3 - 4 | 245 | 75 | |
XM_005002672.3 | XP_005002729.1 | 3 - 4 | 240 | 75 | ||
GenBank mRNA | AJ537436.1 | - | 3 - 4 | 245 | 75 | |
AJ537496.1 | - | 3 - 4 | 245 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
