Assay Details
| Gene Symbol: Bri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172827.1 | NP_001166298.1 | 1 - 2 | 144 | 70 | |
| GenBank mRNA | AB058506.1 | - | 1 - 2 | 144 | 70 | |
More Information
Set Membership: |
|

Bri3
Genomic Map