Assay Details
Gene Symbol: Bfsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173038.1 | NP_001166509.1 | 3 - 4 | 518 | 65 | |
XM_013159547.2 | XP_013015001.2 | 3 - 4 | 707 | 65 | ||
XM_013159549.2 | XP_013015003.1 | 3 - 4 | 460 | 65 | ||
GenBank mRNA | EU827599.1 | - | 3 - 4 | 518 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
