Assay Details
Gene Symbol: Vim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173040.1 | NP_001166511.1 | 1 - 2 | 690 | 73 | |
GenBank mRNA | EU827601.1 | - | 1 - 2 | 690 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
