Assay Details
| Gene Symbol: Nrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173050.1 | NP_001166521.1 | 2 - 3 | 525 | 86 | |
| XM_005005572.3 | XP_005005629.1 | 3 - 4 | 664 | 86 | ||
| GenBank mRNA | EB364156.1 | - | 2 - 3 | 519 | 86 | |
| EU477759.1 | - | 2 - 3 | 525 | 86 | ||
More Information
Set Membership: |
|

Nrl
Genomic Map