Assay Details
Gene Symbol: Ppyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173403.1 | NP_001166874.1 | 1 | 1052 | 65 | |
XM_013148531.2 | XP_013003985.1 | 2 | 1253 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
