Assay Details
| Gene Symbol: Cd1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172744.1 | NP_001166215.1 | 1 - 2 | 64 | 62 | |
| GenBank mRNA | AF145490.1 | - | 1 - 2 | 64 | 62 | |
More Information
Set Membership: |
|

Cd1e
Genomic Map