Assay Details
Gene Symbol: Cd1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172744.1 | NP_001166215.1 | 3 - 4 | 636 | 82 | |
GenBank mRNA | AF145490.1 | - | 3 - 4 | 636 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
