Assay Details
Gene Symbol: Cxcl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173399.2 | NP_001166870.1 | 1 - 2 | 114 | 64 | |
GenBank mRNA | L04986.1 | - | 1 - 2 | 114 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
