Assay Details
Gene Symbol: Gstm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172834.1 | NP_001166305.1 | 7 - 8 | 635 | 74 | |
GenBank mRNA | AB000488.1 | - | 7 - 8 | 635 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
