Assay Details
| Gene Symbol: Gh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172859.1 | NP_001166330.1 | 2 - 3 | 224 | 79 | |
| GenBank mRNA | AF233853.1 | - | 2 - 3 | 224 | 79 | |
| AF238493.1 | - | 1 - 2 | 98 | 79 | ||
More Information
Set Membership: |
|

Gh1
Genomic Map