Assay Details
Gene Symbol: MRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316390.1 | NP_001303319.1 | 10 - 11 | 3835 | 64 | |
NM_135747.4 | NP_609591.2 | 9 - 10 | 3717 | 64 | ||
NM_165020.3 | NP_723772.2 | 9 - 10 | 3724 | 64 | ||
NM_205968.2 | NP_995690.1 | 9 - 10 | 3717 | 64 | ||
NM_205969.2 | NP_995691.1 | 10 - 11 | 3835 | 64 | ||
NM_205970.2 | NP_995692.1 | 10 - 11 | 3835 | 64 | ||
NM_205971.2 | NP_995693.1 | 9 - 10 | 3720 | 64 | ||
NM_205972.2 | NP_995694.1 | 9 - 10 | 3717 | 64 | ||
NM_205973.2 | NP_995695.1 | 10 - 11 | 3835 | 64 | ||
NM_205974.2 | NP_995696.1 | 9 - 10 | 3717 | 64 | ||
NM_205975.2 | NP_995697.1 | 9 - 10 | 3717 | 64 | ||
NM_205976.2 | NP_995698.1 | 9 - 10 | 3717 | 64 | ||
NM_205977.2 | NP_995699.1 | 10 - 11 | 3835 | 64 | ||
NM_205978.2 | NP_995700.1 | 9 - 10 | 3717 | 64 | ||
NM_205979.2 | NP_995701.1 | 10 - 11 | 3835 | 64 | ||
NM_205980.2 | NP_995702.1 | 10 - 11 | 3838 | 64 | ||
NM_205981.2 | NP_995703.1 | 10 - 11 | 3835 | 64 | ||
NM_205982.2 | NP_995704.1 | 10 - 11 | 3835 | 64 | ||
GenBank mRNA | BT150389.2 | - | 9 - 10 | 3696 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
