Assay Details
| Gene Symbol: ACXC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038813.2 | NP_001033902.1 | 4 - 5 | 1255 | 67 | |
| NM_135749.2 | NP_609593.1 | 4 - 5 | 1255 | 67 | ||
| GenBank mRNA | AF177930.1 | - | 4 - 5 | 1256 | 67 | |
| BT021449.1 | - | 4 - 5 | 1242 | 67 | ||
More Information
Set Membership: |
|

ACXC
Genomic Map