Assay Details
Gene Symbol: ACXC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038813.2 | NP_001033902.1 | 7 - 8 | 2570 | 96 | |
NM_135749.2 | NP_609593.1 | 8 - 9 | 2511 | 96 | ||
GenBank mRNA | AF177930.1 | - | 8 - 9 | 2512 | 96 | |
BT021449.1 | - | 8 - 9 | 2498 | 96 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
