Assay Details
| Gene Symbol: CG9934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169496.2 | NP_001162967.1 | 5 - 6 | 3076 | 59 | |
| NM_135753.4 | NP_609597.1 | 5 - 6 | 3366 | 59 | ||
| GenBank mRNA | AY094803.1 | - | 5 - 6 | 2994 | 59 | |
More Information
Set Membership: |
|

CG9934
Genomic Map