Assay Details
Gene Symbol: Sfmbt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144349.2 | NP_001137821.1 | 3 - 4 | 1294 | 60 | |
NM_001273503.1 | NP_001260432.1 | 3 - 4 | 1282 | 60 | ||
NM_135762.3 | NP_609606.2 | 3 - 4 | 1294 | 60 | ||
GenBank mRNA | BT006011.1 | - | 3 - 4 | 1292 | 60 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
