Assay Details
| Gene Symbol: ics | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298973.1 | NP_001285902.1 | 1 - 2 | 162 | 63 | |
| NM_001298974.1 | NP_001285903.1 | 2 - 3 | 743 | 63 | ||
| NM_135821.5 | NP_609665.2 | 3 - 4 | 434 | 63 | ||
| GenBank mRNA | BT031285.1 | - | 1 - 2 | 162 | 63 | |
| BT088923.1 | - | 3 - 4 | 400 | 63 | ||
More Information
Set Membership: |
|

ics
Genomic Map