Assay Details
Gene Symbol: Sec71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135831.3 | NP_609675.2 | 1 - 2 | 1111 | 64 | |
GenBank mRNA | AY122171.1 | - | 1 - 2 | 1082 | 64 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
