Assay Details
Gene Symbol: NimB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201873.2 | NP_001188802.1 | 1 - 2 | 358 | 103 | |
NM_001298990.1 | NP_001285919.1 | 2 - 3 | 418 | 103 | ||
NM_135848.5 | NP_609692.3 | 1 - 2 | 358 | 103 | ||
GenBank mRNA | BT024337.1 | - | 1 - 2 | 337 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
