Assay Details
| Gene Symbol: NimC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135854.3 | NP_609698.2 | 1 | 1081 | 61 | |
| GenBank mRNA | BT030809.1 | - | 1 | 1078 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NimC4| Gene Symbol: NimC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135854.3 | NP_609698.2 | 1 | 1081 | 61 | |
| GenBank mRNA | BT030809.1 | - | 1 | 1078 | 61 | |
Set Membership: |
|