Assay Details
Gene Symbol: Pburs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135868.2 | NP_609712.1 | 1 - 2 | 266 | 62 | |
GenBank mRNA | AY823257.1 | - | 1 - 2 | 253 | 62 | |
BT122094.1 | - | 1 - 2 | 253 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
