Assay Details
Gene Symbol: PRL-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273558.1 | NP_001260487.1 | 4 - 5 | 1675 | 82 | |
NM_001273559.1 | NP_001260488.1 | 4 - 5 | 618 | 82 | ||
NM_135936.3 | NP_609780.1 | 4 - 5 | 1658 | 82 | ||
NM_165150.2 | NP_723956.1 | 4 - 5 | 1292 | 82 | ||
GenBank mRNA | AY071505.1 | - | 4 - 5 | 1662 | 82 |
Gene Symbol: CG46309 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001347787.1 | NP_001334740.1 | 4 - 5 | 1292 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
