Assay Details
Gene Symbol: bur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299191.1 | NP_001286120.1 | 6 - 7 | 1532 | 61 | |
NM_138759.4 | NP_620114.3 | 6 - 7 | 1532 | 61 | ||
GenBank mRNA | AY118572.1 | - | 6 - 7 | 1509 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
