Assay Details
| Gene Symbol: Alp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275230.1 | NP_001262159.1 | 1 - 2 | 143 | 77 | |
| NM_141058.3 | NP_649315.1 | 1 - 2 | 143 | 77 | ||
| GenBank mRNA | AY069739.1 | - | 1 - 2 | 127 | 77 | |
More Information
Set Membership: |
|

Alp1
Genomic Map