Assay Details
| Gene Symbol: Alp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275230.1 | NP_001262159.1 | 2 - 3 | 397 | 76 | |
| NM_141058.3 | NP_649315.1 | 2 - 3 | 397 | 76 | ||
| GenBank mRNA | AY069739.1 | - | 2 - 3 | 381 | 76 | |
More Information
Set Membership: |
|

Alp1
Genomic Map