Assay Details
| Gene Symbol: Est-Q | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_141064.3 | NP_649321.1 | 3 - 4 | 1092 | 61 | |
| GenBank mRNA | AY051599.1 | - | 3 - 4 | 1057 | 61 | |
More Information
Set Membership: |
|

Est-Q
Genomic Map