Assay Details
| Gene Symbol: TfAP-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275249.1 | NP_001262178.1 | 1 - 2 | 482 | 66 | |
| NM_141079.3 | NP_649336.1 | 1 - 2 | 482 | 66 | ||
| GenBank mRNA | AF033030.1 | - | 1 - 2 | 466 | 66 | |
| AY071140.1 | - | 1 - 2 | 480 | 66 | ||
More Information
Set Membership: |
|

TfAP-2
Genomic Map