Assay Details
| Gene Symbol: Alg11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275253.2 | NP_001262182.1 | 3 - 4 | 477 | 81 | |
| NM_141083.3 | NP_649340.1 | 2 - 3 | 301 | 81 | ||
| GenBank mRNA | AY061012.1 | - | 2 - 3 | 288 | 81 | |
| AY113436.1 | - | 2 - 3 | 292 | 81 | ||
More Information
Set Membership: |
|

Alg11
Genomic Map