Assay Details
| Gene Symbol: MCPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169651.2 | NP_001163122.1 | 3 - 4 | 790 | 53 | |
| NM_001169652.2 | NP_001163123.1 | 4 - 5 | 1001 | 53 | ||
| NM_136867.3 | NP_610711.1 | 4 - 5 | 1001 | 53 | ||
| NM_165852.2 | NP_725088.1 | 3 - 4 | 790 | 53 | ||
| GenBank mRNA | AY122187.1 | - | 4 - 5 | 965 | 53 | |
| BT088945.1 | - | 3 - 4 | 797 | 53 | ||
| EF587234.1 | - | 3 - 4 | 790 | 53 | ||
More Information
Set Membership: |
|

MCPH1
Genomic Map