Assay Details
| Gene Symbol: RpS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165868.2 | NP_725114.1 | 2 - 3 | 168 | 63 | |
| GenBank mRNA | KX531537.1 | - | 1 - 2 | 139 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RpS11| Gene Symbol: RpS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165868.2 | NP_725114.1 | 2 - 3 | 168 | 63 | |
| GenBank mRNA | KX531537.1 | - | 1 - 2 | 139 | 63 | |
Set Membership: |
|