Assay Details
Gene Symbol: garz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259349.2 | NP_001246278.1 | 2 - 3 | 614 | 75 | |
NM_136917.4 | NP_610761.2 | 2 - 3 | 614 | 75 | ||
NM_165881.2 | NP_725133.1 | 2 - 3 | 614 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
