Assay Details
Gene Symbol: garz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259349.2 | NP_001246278.1 | 7 - 8 | 4253 | 110 | |
NM_136917.4 | NP_610761.2 | 7 - 8 | 4253 | 110 | ||
NM_165881.2 | NP_725133.1 | 7 - 8 | 4253 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
