Assay Details
| Gene Symbol: Cpr49Ac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169656.2 | NP_001163127.1 | 5 - 6 | 927 | 71 | |
| NM_001299415.1 | NP_001286344.1 | 5 - 6 | 511 | 71 | ||
| NM_001299416.1 | NP_001286345.1 | 5 - 6 | 601 | 71 | ||
| NM_001299417.1 | NP_001286346.1 | 5 - 6 | 895 | 71 | ||
| NM_136928.5 | NP_610772.3 | 6 - 7 | 538 | 71 | ||
| NM_165892.4 | NP_725150.2 | 6 - 7 | 628 | 71 | ||
| GenBank mRNA | AY052054.1 | - | 6 - 7 | 519 | 71 | |
| BT003229.1 | - | 6 - 7 | 609 | 71 | ||
More Information
Set Membership: |
|

Cpr49Ac
Genomic Map