Assay Details
Gene Symbol: fra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299424.1 | NP_001286353.1 | 8 - 9 | 4866 | 59 | |
NM_078992.3 | NP_523716.2 | 8 - 9 | 4893 | 59 | ||
NM_165913.2 | NP_725185.1 | 8 - 9 | 4440 | 59 | ||
GenBank mRNA | BT150366.1 | - | 8 - 9 | 4890 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
