Assay Details
| Gene Symbol: Dgkepsilon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299433.1 | NP_001286362.1 | 1 - 2 | 280 | 69 | |
| NM_165949.2 | NP_725228.1 | 1 - 2 | 364 | 69 | ||
| GenBank mRNA | AY050241.1 | - | 1 - 2 | 247 | 69 | |
More Information
Set Membership: |
|

Dgkepsilon
Genomic Map