Assay Details
| Gene Symbol: NAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038854.2 | NP_001033943.2 | 3 - 4 | 4765 | 101 | |
| NM_143770.3 | NP_652027.2 | 3 - 4 | 4741 | 101 | ||
| NM_165961.2 | NP_725242.1 | 3 - 4 | 4765 | 101 | ||
| GenBank mRNA | BT150476.1 | - | 3 - 4 | 4762 | 101 | |
More Information
Set Membership: |
|

NAT1
Genomic Map