Assay Details
Gene Symbol: cnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014523.2 | NP_001014523.1 | 1 - 2 | 162 | 63 | |
NM_001169670.2 | NP_001163141.1 | 1 - 2 | 162 | 63 | ||
NM_001259362.1 | NP_001246291.1 | 1 - 2 | 162 | 63 | ||
NM_001259367.1 | NP_001246296.1 | 1 - 2 | 162 | 63 | ||
NM_165991.4 | NP_725298.1 | 1 - 2 | 162 | 63 | ||
GenBank mRNA | BT001287.1 | - | 1 - 2 | 150 | 63 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
