Assay Details
| Gene Symbol: cnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169670.2 | NP_001163141.1 | 3 - 4 | 808 | 77 | |
| NM_001259362.1 | NP_001246291.1 | 3 - 4 | 808 | 77 | ||
| NM_001259363.2 | NP_001246292.1 | 4 - 5 | 990 | 77 | ||
| NM_001259364.2 | NP_001246293.1 | 4 - 5 | 990 | 77 | ||
| NM_001259367.1 | NP_001246296.1 | 3 - 4 | 808 | 77 | ||
| NM_165991.4 | NP_725298.1 | 3 - 4 | 808 | 77 | ||
| GenBank mRNA | BT001287.1 | - | 3 - 4 | 796 | 77 | |
More Information
Set Membership: |
|

cnn
Genomic Map