Assay Details
| Gene Symbol: cnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259364.2 | NP_001246293.1 | 8 - 9 | 1677 | 74 | |
| NM_165991.4 | NP_725298.1 | 7 - 8 | 1495 | 74 | ||
More Information
Set Membership: |
|

cnn
Genomic Map